missing translation for 'onlineSavingsMsg'
Learn More

(4-Bromothien-3-yl)methanol, 97%, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals CC54209DA

Quantity 1g
Packaging Amber Glass Bottle

 View more versions of this product

Catalog No. CC54209DA

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

Specifications

Chemical Identifiers

70260-05-8
193.06
MPKBPMYJCDNVEF-UHFFFAOYSA-N
818863
OCC1=CSC=C1Br
C5H5BrOS
MFCD01859831
4-bromothiophen-3-yl methanol, 4-bromothien-3-yl methanol, 4-bromo-3-thiophenemethanol, 3-thiophenemethanol,4-bromo, 4-bromothiophen-3-yl ;methanol, 4-bromo-3-thienyl methan-1-ol, 3-bromo-4-hydroxymethyl thiophene
(4-bromothiophen-3-yl)methanol

Specifications

(4-Bromothien-3-yl)methanol
96.5
97%
C5H5BrOS
MFCD01859831
MPKBPMYJCDNVEF-UHFFFAOYSA-N
(4-bromothiophen-3-yl)methanol
818863
97%
70260-05-8
100.0
Amber Glass Bottle
1g
4-bromothiophen-3-yl methanol, 4-bromothien-3-yl methanol, 4-bromo-3-thiophenemethanol, 3-thiophenemethanol,4-bromo, 4-bromothiophen-3-yl ;methanol, 4-bromo-3-thienyl methan-1-ol, 3-bromo-4-hydroxymethyl thiophene
OCC1=CSC=C1Br
193.06
193.06
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Harmful if swallowed.
Causes serious eye irritation.
Harmful if inhaled.
Harmful in contact with skin.
Causes skin irritation.
May cause respiratory irritation.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

SDS
Documents

Documents