missing translation for 'onlineSavingsMsg'
Learn More

4-Bromopyridine hydrochloride, 99%

Catalog No. p-7044547
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0705206 5 g
AAL0705214 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0705206 Supplier Thermo Scientific Chemicals Supplier No. L0705206

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 19524-06-2 | C5H5BrClN | 194.456 g/mol

4-Bromopyridine hydrochloride is used as a useful synthetic intermediate. It is also used as an important raw material and intermediate used in Organic Synthesis, Pharmaceuticals, Agrochemicals and Dyestuff.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Bromopyridine hydrochloride is used as a useful synthetic intermediate. It is also used as an important raw material and intermediate used in Organic Synthesis, Pharmaceuticals, Agrochemicals and Dyestuff.

Solubility
Soluble in DMSO, Methanol and Water.

Notes
Store at -20°C. Hygroscopic. Incompatible materials are moisture, strong bases, strong oxidizing agents. Protect from humidity and air.

Chemical Identifiers

CAS 19524-06-2
Molecular Formula C5H5BrClN
Molecular Weight (g/mol) 194.456
MDL Number MFCD00012828
InChI Key MPZMVUQGXAOJIK-UHFFFAOYSA-N
Synonym 4-bromopyridine hydrochloride, 4-bromopyridinium chloride, 4-bromopyridine hcl, pyridine, 4-bromo-, hydrochloride, 4-bromo-pyridine hcl, 4-bromo pyridine hcl, 4-bromopyridine, chloride, 4-bromopyridinehydrochloride, 4-bromopyridine hydrochloride salt, 4-bromopyridine-hcl
PubChem CID 88100
IUPAC Name 4-bromopyridine;hydrochloride
SMILES C1=CN=CC=C1Br.Cl

Specifications

Melting Point ∽270°C (decomposition)
Quantity 5 g
Beilstein 3621847
Sensitivity Hygroscopic
Solubility Information Soluble in DMSO,Methanol and Water.
Formula Weight 194.46
Percent Purity 99%
Chemical Name or Material 4-Bromopyridine hydrochloride
Hazard Category H302+H312+H332-H315-H319-H335
Hazard Statement GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
EINECSNumber 243-128-4
TSCA No
Recommended Storage Ambient temperatures; Store under Nitrogen

RUO – Research Use Only