Learn More
4-Bromopyridine Hydrochloride, 98%
CAS: 19524-06-2 | C5H4BrN·HCl | 194.46 g/mol
$221.05 - $221.05
Chemical Identifiers
| CAS | 19524-06-2 |
|---|---|
| Molecular Formula | C5H4BrN·HCl |
| Molecular Weight (g/mol) | 194.46 |
| MDL Number | MFCD00012828 |
| InChI Key | MPZMVUQGXAOJIK-UHFFFAOYSA-N |
| Synonym | 4-bromopyridine hydrochloride, 4-bromopyridinium chloride, 4-bromopyridine hcl, pyridine, 4-bromo-, hydrochloride, 4-bromo-pyridine hcl, 4-bromo pyridine hcl, 4-bromopyridine, chloride, 4-bromopyridinehydrochloride, 4-bromopyridine hydrochloride salt, 4-bromopyridine-hcl |
| PubChem CID | 88100 |
| IUPAC Name | 4-bromopyridine;hydrochloride |
| SMILES | C1=CN=CC=C1Br.Cl |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
AC107420050
|
Thermo Scientific Chemicals
107420050 |
5 g | Glass bottle |
N/A
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 19524-06-2 | |
| 194.46 | |
| MPZMVUQGXAOJIK-UHFFFAOYSA-N | |
| 88100 | |
| C1=CN=CC=C1Br.Cl |
| C5H4BrN·HCl | |
| MFCD00012828 | |
| 4-bromopyridine hydrochloride, 4-bromopyridinium chloride, 4-bromopyridine hcl, pyridine, 4-bromo-, hydrochloride, 4-bromo-pyridine hcl, 4-bromo pyridine hcl, 4-bromopyridine, chloride, 4-bromopyridinehydrochloride, 4-bromopyridine hydrochloride salt, 4-bromopyridine-hcl | |
| 4-bromopyridine;hydrochloride |
Specifications
| 19524-06-2 | |
| 100.0 | |
| Peach to White | |
| 98% | |
| C5H4BrN·HCl | |
| MFCD00012828 | |
| 4-bromopyridine hydrochloride, 4-bromopyridinium chloride, 4-bromopyridine hcl, pyridine, 4-bromo-, hydrochloride, 4-bromo-pyridine hcl, 4-bromo pyridine hcl, 4-bromopyridine, chloride, 4-bromopyridinehydrochloride, 4-bromopyridine hydrochloride salt, 4-bromopyridine-hcl | |
| MPZMVUQGXAOJIK-UHFFFAOYSA-N | |
| 4-bromopyridine;hydrochloride | |
| 88100 | |
| 98% | |
| 4-Bromopyridine hydrochloride |
| 97.5 | |
| 313.0°C to 315.0°C | |
| Authentic | |
| Glass bottle | |
| (C5H4N)Br·HCl | |
| 5 g | |
| Solubility in water: soluble | |
| C1=CN=CC=C1Br.Cl | |
| 194.46 | |
| 194.46 | |
| Powder |
Safety and Handling
GHS H Statement
Harmful if inhaled.
Causes serious eye irritation.
May cause respiratory irritation.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Call a POISON CENTER or doctor/physicia
GHS Signal Word: Warning
EINECSNumber : 243-128-4
RUO – Research Use Only