Learn More
4-Bromopyridine-2-carboxylic acid, 97%
CAS: 30766-03-1 | C6H4BrNO2 | 202.007 g/mol
Supplier: Thermo Scientific Chemicals H6424414
Description
4-Bromopicolinic acid is a useful synthetic intermediate. Also used as a intermediate for agrochemical, pharmaceutical and dyestuff field.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications4-Bromopicolinic acid is a useful synthetic intermediate. Also used as a intermediate for agrochemical, pharmaceutical and dyestuff field.
Solubility
Sparingly soluble in dimethyl sulfoxide , methanol and water.
Notes
Store in cool, dry place in tightly closed container. With adequate ventilation. Store away from oxidizing agent.
Specifications
| 4-Bromopyridine-2-carboxylic acid | |
| 177°C to 179°C | |
| MFCD03426665 | |
| 471891 | |
| Sparingly soluble in dimethyl sulfoxide ,methanol and water. | |
| C1=CN=C(C=C1Br)C(=O)O | |
| 202.007 | |
| 202.01 |
| 30766-03-1 | |
| C6H4BrNO2 | |
| 25 g | |
| 4-bromopicolinic acid, 4-bromo-2-pyridinecarboxylic acid, 4-bromo-pyridinecarboxylic acid, 2-pyridinecarboxylic acid, 4-bromo, 4-bromo-pyridine-2-carboxylic acid, aurora 23242, pubchem4064, acmc-209hio, ksc222g5r | |
| RPHHYRNGCJYQSP-UHFFFAOYSA-N | |
| 4-bromopyridine-2-carboxylic acid | |
| 735150 | |
| 97% |
Chemical Identifiers
| 30766-03-1 | |
| 202.007 | |
| RPHHYRNGCJYQSP-UHFFFAOYSA-N | |
| 735150 | |
| C1=CN=C(C=C1Br)C(=O)O |
| C6H4BrNO2 | |
| MFCD03426665 | |
| 4-bromopicolinic acid, 4-bromo-2-pyridinecarboxylic acid, 4-bromo-pyridinecarboxylic acid, 2-pyridinecarboxylic acid, 4-bromo, 4-bromo-pyridine-2-carboxylic acid, aurora 23242, pubchem4064, acmc-209hio, ksc222g5r | |
| 4-bromopyridine-2-carboxylic acid |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only