missing translation for 'onlineSavingsMsg'
Learn More

4-Bromopyridine-2-carboxylic acid, 97%

CAS: 30766-03-1 | C6H4BrNO2 | 202.007 g/mol

Supplier:  Thermo Scientific Chemicals H6424406

 View more versions of this product

Catalog No. AAH6424406


Only null left
Explore available promotions

Description

Description

4-Bromopicolinic acid is a useful synthetic intermediate. Also used as a intermediate for agrochemical, pharmaceutical and dyestuff field.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Bromopicolinic acid is a useful synthetic intermediate. Also used as a intermediate for agrochemical, pharmaceutical and dyestuff field.

Solubility
Sparingly soluble in dimethyl sulfoxide , methanol and water.

Notes
Store in cool, dry place in tightly closed container. With adequate ventilation. Store away from oxidizing agent.
Specifications

Specifications

4-Bromopyridine-2-carboxylic acid
177°C to 179°C
MFCD03426665
471891
Sparingly soluble in dimethyl sulfoxide ,methanol and water.
C1=CN=C(C=C1Br)C(=O)O
202.007
202.01
30766-03-1
C6H4BrNO2
5 g
4-bromopicolinic acid, 4-bromo-2-pyridinecarboxylic acid, 4-bromo-pyridinecarboxylic acid, 2-pyridinecarboxylic acid, 4-bromo, 4-bromo-pyridine-2-carboxylic acid, aurora 23242, pubchem4064, acmc-209hio, ksc222g5r
RPHHYRNGCJYQSP-UHFFFAOYSA-N
4-bromopyridine-2-carboxylic acid
735150
97%

Chemical Identifiers

30766-03-1
202.007
RPHHYRNGCJYQSP-UHFFFAOYSA-N
735150
C1=CN=C(C=C1Br)C(=O)O
C6H4BrNO2
MFCD03426665
4-bromopicolinic acid, 4-bromo-2-pyridinecarboxylic acid, 4-bromo-pyridinecarboxylic acid, 2-pyridinecarboxylic acid, 4-bromo, 4-bromo-pyridine-2-carboxylic acid, aurora 23242, pubchem4064, acmc-209hio, ksc222g5r
4-bromopyridine-2-carboxylic acid
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only