missing translation for 'onlineSavingsMsg'
Learn More

4-Bromopyrazole 98.0+%, TCI America™

Catalog No. p-7123139
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B37455G 5 g
B374525G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B37455G Supplier TCI America Supplier No. B37455G

Chemical Identifiers

CAS 2075-45-8
Molecular Formula C3H3BrN2
Molecular Weight (g/mol) 146.98
MDL Number MFCD00075602
InChI Key WVGCPEDBFHEHEZ-UHFFFAOYSA-N
PubChem CID 16375
IUPAC Name 4-bromo-1H-pyrazole
SMILES BrC1=CNN=C1

Specifications

Melting Point 97°C
Color White-Yellow
Boiling Point 260°C
Quantity 5 g
Synonym 4-bromopyrazole, 1h-pyrazole, 4-bromo, 4-brompyrazole, 4-bromo-pyrazole, pyrazole, 4-bromo, 4-bromo pyrazole, byz, zlchem 126, 1h-pyrazole,4-bromo, pubchem11041
Formula Weight 146.98
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4-Bromopyrazole
RTECSNumber UQ6200000
TSCA No