missing translation for 'onlineSavingsMsg'
Learn More

4-Bromopyrazole, 98%

Catalog No. AC396820010
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Packaging:
Glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC396820010 1 g Glass bottle
AC396820050 5 g Glass bottle
AC396820250 25 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC396820010 Supplier Thermo Scientific Chemicals Supplier No. 396820010
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 2075-45-8 | C3H3BrN2 | 146.98 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 2075-45-8
Molecular Formula C3H3BrN2
Molecular Weight (g/mol) 146.98
MDL Number MFCD00075602
InChI Key WVGCPEDBFHEHEZ-UHFFFAOYSA-N
PubChem CID 16375
IUPAC Name 4-bromo-1H-pyrazole
SMILES BrC1=CNN=C1

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 93.0°C to 96.0°C
Color White to Cream
Boiling Point 250.0°C to 260.0°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (HPLC)
Packaging Glass bottle
Quantity 1 g
Synonym 4-bromopyrazole, 1h-pyrazole, 4-bromo, 4-brompyrazole, 4-bromo-pyrazole, pyrazole, 4-bromo, 4-bromo pyrazole, byz, zlchem 126, 1h-pyrazole,4-bromo, pubchem11041
Formula Weight 146.97
Percent Purity 98%
Physical Form Crystalline Powder
Chemical Name or Material 4-Bromopyrazole
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

RUO – Research Use Only