missing translation for 'onlineSavingsMsg'
Learn More

4-Bromopiperidine Hydrobromide 98.0+%, TCI America™

Catalog No. B38301G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B38301G 1 g
B38305G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B38301G Supplier TCI America Supplier No. B38301G

Chemical Identifiers

CAS 54288-70-9
Molecular Formula C5H11Br2N
Molecular Weight (g/mol) 244.958
MDL Number MFCD00191858
InChI Key LVTIZXGNLIKUQZ-UHFFFAOYSA-N
Synonym 4-bromopiperidine hydrobromide, 4-bromopiperidinehbr, 4-bromopiperidine hbr, piperidine, 4-bromo-, hydrobromide, 4-bromopiperidine hydrobromide 1:1, 4-bromopiperidine, bromide, pubchem9060, acmc-209lf5, 4-bromo-piperidine hbr salt, 4-bromopiperidinehydrobromide
PubChem CID 2734676
IUPAC Name 4-bromopiperidine;hydrobromide
SMILES C1CNCCC1Br.Br

Specifications

Melting Point 192°C
Color White
Quantity 1 g
Formula Weight 244.96
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 4-Bromopiperidine Hydrobromide