missing translation for 'onlineSavingsMsg'
Learn More

4-Bromopiperidine hydrobromide, 98%, Thermo Scientific™

Catalog No. p-2690608
Change view
Click to view available options
Quantity:
1g
10g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC368310010 1g Glass bottle
AC368310100 10g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC368310010 Supplier Thermo Scientific Chemicals Supplier No. 368310010

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Chemical Identifiers

CAS 54288-70-9
Molecular Formula C5H10BrN·HBr
Molecular Weight (g/mol) 244.96
MDL Number MFCD00191858
InChI Key LVTIZXGNLIKUQZ-UHFFFAOYSA-N
Synonym 4-bromopiperidine hydrobromide, 4-bromopiperidinehbr, 4-bromopiperidine hbr, piperidine, 4-bromo-, hydrobromide, 4-bromopiperidine hydrobromide 1:1, 4-bromopiperidine, bromide, pubchem9060, acmc-209lf5, 4-bromo-piperidine hbr salt, 4-bromopiperidinehydrobromide
PubChem CID 2734676
IUPAC Name 4-bromopiperidine;hydrobromide
SMILES C1CNCCC1Br.Br

Specifications

CAS Min % 97.5
CAS Max % 100.0
Infrared Spectrum Authentic
Assay Percent Range 97.5 to 102.5% (Argentometry)
Packaging Glass bottle
Quantity 1g
Beilstein 20, IV, 1381
Formula Weight 244.96
Percent Purity 98%
Chemical Name or Material 4-Bromopiperidine hydrobromide
Hazard Statement GHS H Statement:
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

RUO – Research Use Only