missing translation for 'onlineSavingsMsg'
Learn More

4-Bromophenylacetic acid, 99%, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals 160521000

Quantity 100g
Packaging Glass bottle

 View more versions of this product

Catalog No. AC160521000


Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

1878-68-8
215.05
QOWSWEBLNVACCL-UHFFFAOYSA-N
74654
2-(4-bromophenyl)acetic acid
C8H7BrO2
MFCD00004342
4-bromophenylacetic acid, 2-4-bromophenyl acetic acid, 4-bromophenyl acetic acid, p-bromophenylacetic acid, 4-bromobenzeneacetic acid, benzeneacetic acid, 4-bromo, p-bromophenyl acetic acid, acetic acid, p-bromophenyl, p-bromphenylessigsaure german
CHEBI:1790
OC(=O)CC1=CC=C(Br)C=C1

Specifications

4-Bromophenylacetic acid
98.5
Authentic
Glass bottle
BrC6H4CH2CO2H
MFCD00004342
4-bromophenylacetic acid, 2-4-bromophenyl acetic acid, 4-bromophenyl acetic acid, p-bromophenylacetic acid, 4-bromobenzeneacetic acid, benzeneacetic acid, 4-bromo, p-bromophenyl acetic acid, acetic acid, p-bromophenyl, p-bromphenylessigsaure german
QOWSWEBLNVACCL-UHFFFAOYSA-N
2-(4-bromophenyl)acetic acid
74654
215.05
1878-68-8
100.0
99%
C8H7BrO2
100g
09, 451
Solubility in water: slightly soluble in water. Other solubilities: very soluble in ethyl alcohol,diethyl ether,carb
OC(=O)CC1=CC=C(Br)C=C1
215.05
CHEBI:1790
99%
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 217-523-7

SDS
Documents

Documents

Promotions

Promotions