missing translation for 'onlineSavingsMsg'
Learn More

4-Bromophenylacetic acid, 99%, Thermo Scientific™

Catalog No. AC160520250
Change view
Click to view available options
Quantity:
25g
100g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC160520250 25g Glass bottle
AC160521000 100g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC160520250 Supplier Thermo Scientific Chemicals Supplier No. 160520250

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Chemical Identifiers

CAS 1878-68-8
Molecular Formula C8H7BrO2
Molecular Weight (g/mol) 215.05
MDL Number MFCD00004342
InChI Key QOWSWEBLNVACCL-UHFFFAOYSA-N
Synonym 4-bromophenylacetic acid, 2-4-bromophenyl acetic acid, 4-bromophenyl acetic acid, p-bromophenylacetic acid, 4-bromobenzeneacetic acid, benzeneacetic acid, 4-bromo, p-bromophenyl acetic acid, acetic acid, p-bromophenyl, p-bromphenylessigsaure german
PubChem CID 74654
ChEBI CHEBI:1790
IUPAC Name 2-(4-bromophenyl)acetic acid
SMILES OC(=O)CC1=CC=C(Br)C=C1

Specifications

CAS Min % 98.5
CAS Max % 100.0
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Linear Formula BrC6H4CH2CO2H
Quantity 25g
Beilstein 09, 451
Solubility Information Solubility in water: slightly soluble in water. Other solubilities: very soluble in ethyl alcohol,diethyl ether,carb
Formula Weight 215.05
Percent Purity 99%
Chemical Name or Material 4-Bromophenylacetic acid
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 217-523-7

RUO – Research Use Only