missing translation for 'onlineSavingsMsg'
Learn More

(4-Bromophenoxy)-tert-butyldimethylsilane, 97%, Thermo Scientific™

Chemical Identifiers

CAS 67963-68-2
Molecular Formula C12H19BrOSi
Molecular Weight (g/mol) 287.27
MDL Number MFCD00239382
InChI Key DLGZGLKSNRKLSM-UHFFFAOYSA-N
Synonym 4-bromophenoxy tert-butyl dimethylsilane, 4-bromophenoxy-tert-butyldimethylsilane, p-bromophenoxy t-butyl dimethylsilane, 4-bromophenoxy-tert-butyl-dimethylsilane, 1-bromo-4-tert-butyldimethylsiloxy benzene, 4-bromophenol tert-butyldimethylsilyl ether, silane, 4-bromophenoxy 1,1-dimethylethyl dimethyl, 1-bromo-4-1,1-dimethylethyl dimethylsilyl oxy benzene, acmc-1b8oo
PubChem CID 4227167
IUPAC Name (4-bromophenoxy)-tert-butyl-dimethylsilane
SMILES CC(C)(C)[Si](C)(C)OC1=CC=C(C=C1)Br
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC427090050
SDS View Documents
Thermo Scientific Chemicals
427090050
5mL Glass bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AC427090250
SDS View Documents
Thermo Scientific Chemicals
427090250
25mL Glass bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

67963-68-2
287.27
DLGZGLKSNRKLSM-UHFFFAOYSA-N
4227167
CC(C)(C)[Si](C)(C)OC1=CC=C(C=C1)Br
C12H19BrOSi
MFCD00239382
4-bromophenoxy tert-butyl dimethylsilane, 4-bromophenoxy-tert-butyldimethylsilane, p-bromophenoxy t-butyl dimethylsilane, 4-bromophenoxy-tert-butyl-dimethylsilane, 1-bromo-4-tert-butyldimethylsiloxy benzene, 4-bromophenol tert-butyldimethylsilyl ether, silane, 4-bromophenoxy 1,1-dimethylethyl dimethyl, 1-bromo-4-1,1-dimethylethyl dimethylsilyl oxy benzene, acmc-1b8oo
(4-bromophenoxy)-tert-butyl-dimethylsilane

Specifications

67963-68-2
100.0
137°C (25.0mmHg)
96% min. (GC)
C12H19BrOSi
MFCD00239382
1.174
DLGZGLKSNRKLSM-UHFFFAOYSA-N
(4-bromophenoxy)-tert-butyl-dimethylsilane
4227167
97%
96.0
1.1740g/mL
12°C
Glass bottle
BrC6H4OSi(CH3)2C(CH3)3
5mL
4-bromophenoxy tert-butyl dimethylsilane, 4-bromophenoxy-tert-butyldimethylsilane, p-bromophenoxy t-butyl dimethylsilane, 4-bromophenoxy-tert-butyl-dimethylsilane, 1-bromo-4-tert-butyldimethylsiloxy benzene, 4-bromophenol tert-butyldimethylsilyl ether, silane, 4-bromophenoxy 1,1-dimethylethyl dimethyl, 1-bromo-4-1,1-dimethylethyl dimethylsilyl oxy benzene, acmc-1b8oo
CC(C)(C)[Si](C)(C)OC1=CC=C(C=C1)Br
287.27
287.27
(4-Bromophenoxy)-tert-butyldimethylsilane
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.
Highly flammable liquid and vapor.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only