missing translation for 'onlineSavingsMsg'
Learn More

4-Bromophenethylamine, 98%, Thermo Scientific Chemicals

Catalog No. AC183090010
Change view
Click to view available options
Quantity:
1 mL
5 mL
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC183090010 1 mL Glass bottle
AC183090050 5 mL Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC183090010 Supplier Thermo Scientific Chemicals Supplier No. 183090010
Get Up to $1,800 in Chemicals with Purchase
View all promotions

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 73918-56-6
Molecular Formula C8H11BrN
Molecular Weight (g/mol) 201.09
MDL Number MFCD00008189
InChI Key ZSZCXAOQVBEPME-UHFFFAOYSA-O
Synonym 4-bromophenethylamine, 2-4-bromophenyl ethylamine, 2-4-bromophenyl ethanamine, p-bromophenethylamine, 2-4-bromo-phenyl-ethylamine, benzeneethanamine, 4-bromo, 2-4-bromophenyl ethan-1-amine, 4-bromophenylethylamine, 4-bromobenzeneethanamine, 1-amino-2-4-bromophenyl ethane
PubChem CID 533915
SMILES [NH3+]CCC1=CC=C(Br)C=C1

Specifications

Color Colorless to Yellow
Density 1.2900g/mL
Boiling Point 63°C to 72°C (0.2 mmHg)
Flash Point >110°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Refractive Index 1.5730 to 1.575
Linear Formula BrC6H4CH2CH2NH2
Quantity 1 mL
Specific Gravity 1.29
IUPAC Name 2-(4-bromophenyl)ethan-1-aminium
Formula Weight 200.08
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material 4-Bromophenethylamine
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement:
Causes serious eye damage.
Causes severe skin burns and eye damage.
Precautionary Statement GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 277-636-2

RUO – Research Use Only