missing translation for 'onlineSavingsMsg'
Learn More

4-(Bromomethyl)pyridine Hydrobromide 97.0+%, TCI America™

Catalog No. p-7122869
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B33155G 5 g
B331525G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B33155G Supplier TCI America Supplier No. B33155G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 73870-24-3
Molecular Formula C6H7Br2N
Molecular Weight (g/mol) 252.94
MDL Number MFCD01863545
InChI Key VAJUUDUWDNCECT-UHFFFAOYSA-N
Synonym 4-bromomethyl pyridine hydrobromide, 4-bromomethylpyridine hydrobromide, 4-bromomethyl-pyridine hydrobromide, 4-pyridylmethyl bromide hydrobromide, pyridine, 4-bromomethyl-, hydrobromide, 4-bromomethyl pyridine, bromide, 4-bromomethyl-pyridine, 4-bromomethyl pyridinium bromide, acmc-1bbrw
PubChem CID 12752219
IUPAC Name hydrogen 4-(bromomethyl)pyridine bromide
SMILES [H+].[Br-].BrCC1=CC=NC=C1

Specifications

Melting Point 186°C
Color White-Yellow
Quantity 5 g
UN Number 3261
Formula Weight 252.94
Percent Purity ≥97.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 4-(Bromomethyl)pyridine Hydrobromide