missing translation for 'onlineSavingsMsg'
Learn More

4-(Bromomethyl)pyridine hydrobromide, 97%

Catalog No. AC397330250
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC397330250 25 g
AC397330050 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC397330250 Supplier Thermo Scientific Chemicals Supplier No. 397330250

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 73870-24-3 | C6H7Br2N | 252.94 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 73870-24-3
Molecular Formula C6H7Br2N
Molecular Weight (g/mol) 252.94
MDL Number MFCD01863545
InChI Key VAJUUDUWDNCECT-UHFFFAOYSA-N
Synonym 4-bromomethyl pyridine hydrobromide, 4-bromomethylpyridine hydrobromide, 4-bromomethyl-pyridine hydrobromide, 4-pyridylmethyl bromide hydrobromide, pyridine, 4-bromomethyl-, hydrobromide, 4-bromomethyl pyridine, bromide, 4-bromomethyl-pyridine, 4-bromomethyl pyridinium bromide, acmc-1bbrw
PubChem CID 12752219
IUPAC Name 4-(bromomethyl)pyridine;hydrobromide
SMILES [H+].[Br-].BrCC1=CC=NC=C1

Specifications

Melting Point 189.0°C to 192.0°C
Assay Percent Range 96% min. (Argentometry)
Packaging Glass bottle
Quantity 25 g
Formula Weight 252.93
Percent Purity 97%
Chemical Name or Material 4-(Bromomethyl)pyridine hydrobromide
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Causes severe skin burns and eye damage.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.

RUO – Research Use Only