missing translation for 'onlineSavingsMsg'
Learn More

4-(Bromomethyl)phenylacetic acid, 97%, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals 392900050

Quantity 5g
Packaging Glass bottle

 View more versions of this product

Catalog No. AC392900050

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

13737-36-5
229.07
WCOCCXZFEJGHTC-UHFFFAOYSA-N
4519056
C1=CC(=CC=C1CC(=O)O)CBr
C9H9BrO2
MFCD00010632
4-bromomethyl phenylacetic acid, 4-bromomethylphenylacetic acid, 2-4-bromomethyl phenyl acetic acid, pam acid, br-pam-linker, 4-carboxymethyl benzyl bromide, 4-bromomethyl phenyl acetic acid, 4-bromomethyl phenylaceticacid
2-[4-(bromomethyl)phenyl]acetic acid

Specifications

4-(Bromomethyl)phenylacetic acid
96.0
Authentic
Glass bottle
BrCH2C6H4CH2COOH
MFCD00010632
WCOCCXZFEJGHTC-UHFFFAOYSA-N
2-[4-(bromomethyl)phenyl]acetic acid
4519056
97%
13737-36-5
100.0
96% min. (HPLC)
C9H9BrO2
5g
4-bromomethyl phenylacetic acid, 4-bromomethylphenylacetic acid, 2-4-bromomethyl phenyl acetic acid, pam acid, br-pam-linker, 4-carboxymethyl benzyl bromide, 4-bromomethyl phenyl acetic acid, 4-bromomethyl phenylaceticacid
C1=CC(=CC=C1CC(=O)O)CBr
229.07
229.07
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Harmful if inhaled.
Causes serious eye irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

SDS
Documents

Documents