missing translation for 'onlineSavingsMsg'
Learn More

4-Bromomethyl-6,7-dimethoxycoumarin 98.0+%, TCI America™

Catalog No. A5570100MG
Change view
Click to view available options
Quantity:
100 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A5570100MG 100 mg
A55701G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A5570100MG Supplier TCI America Supplier No. A5570100MG

Chemical Identifiers

CAS 88404-25-5
Molecular Formula C12H11BrO4
Molecular Weight (g/mol) 299.12
MDL Number MFCD00011570
InChI Key JGODLBJJCNQFII-UHFFFAOYSA-N
Synonym 4-bromomethyl-6,7-dimethoxycoumarin, 4-bromomethyl-6,7-dimethoxy-2h-chromen-2-one, 4-bromomethyl-6,7-dimethoxychromen-2-one, 2h-1-benzopyran-2-one, 4-bromomethyl-6,7-dimethoxy, 4-bromomethyl-6,7-dimethoxy coumarin, 4-bdmc, brdmc-4,6,7, 4-bromomethyl-6,7-dimethoxy-2-oxo-2h-benzopyran
PubChem CID 128870
IUPAC Name 4-(bromomethyl)-6,7-dimethoxychromen-2-one
SMILES COC1=C(C=C2C(=C1)C(=CC(=O)O2)CBr)OC

Specifications

Melting Point 214°C
Quantity 100 mg
UN Number 3261
Formula Weight 299.12
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material 4-Bromomethyl-6,7-dimethoxycoumarin [for HPLC Labeling]
TSCA No
Recommended Storage Refrigerator