missing translation for 'onlineSavingsMsg'
Learn More

4-(Bromomethyl)-5-methyl-3-phenylisoxazole, 97%, Thermo Scientific™

Catalog No. CC00508CB
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
CC00508CB 250 mg
CC00508DA 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. CC00508CB Supplier Thermo Scientific Chemicals Supplier No. CC00508CB

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 180597-83-5
Molecular Formula C11H10BrNO
Molecular Weight (g/mol) 252.11
MDL Number MFCD02677680
InChI Key UICMWXWMCOJBIQ-UHFFFAOYSA-N
PubChem CID 2776147
IUPAC Name 4-(bromomethyl)-5-methyl-3-phenyl-1,2-oxazole
SMILES CC1=C(CBr)C(=NO1)C1=CC=CC=C1

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 250 mg
Synonym 4-bromomethyl-5-methyl-3-phenylisoxazole, 4-bromomethyl-5-methyl-3-phenyl-1,2-oxazole, isoxazole, 4-bromomethyl-5-methyl-3-phenyl, 4-bromomethyl-5-methyl-3-phenylisoxazol, isoxazole,4-bromomethyl-5-methyl-3-phenyl
Formula Weight 252.11
Percent Purity 97%
Chemical Name or Material 4-(Bromomethyl)-5-methyl-3-phenylisoxazole
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement:
Causes serious eye damage.
Causes severe skin burns and eye damage.
Precautionary Statement GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.