missing translation for 'onlineSavingsMsg'
Learn More

4-(Bromomethyl)-4'-methyl-2,2'-bipyridyl 97.0+%, TCI America™

Catalog No. p-7123285
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B3986200MG 200 mg
B39861G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B3986200MG Supplier TCI America Supplier No. B3986200MG

Chemical Identifiers

CAS 81998-05-2
Molecular Formula C12H11BrN2
Molecular Weight (g/mol) 263.138
MDL Number MFCD08275265
InChI Key HUJZHWSADZZJAB-UHFFFAOYSA-N
Synonym 4-(Bromomethyl)-4′C-methyl-2,2′C-bipyridine
PubChem CID 10956379
IUPAC Name 2-[4-(bromomethyl)pyridin-2-yl]-4-methylpyridine
SMILES CC1=CC(=NC=C1)C2=NC=CC(=C2)CBr

Specifications

Melting Point 76°C
Color White-Yellow
Quantity 200 mg
UN Number 3261
Formula Weight 263.14
Percent Purity ≥97.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4-(Bromomethyl)-4′-methyl-2,2′-bipyridyl