missing translation for 'onlineSavingsMsg'
Learn More

4-Bromomethyl-2(1H)-quinolinone, Thermo Scientific Chemicals

$114.74 - $114.74

Chemical Identifiers

CAS 4876-10-2
MDL Number MFCD03426152
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAB2530506
View Documents
Thermo Scientific Chemicals
B2530506
5 g
Each for $114.74
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

4876-10-2
MFCD03426152

Specifications

4876-10-2
C2H4Cl2O
5 g
dichloromethyl methyl ether, 1,1-dichlorodimethyl ether, dichloro methoxy methane, methane, dichloromethoxy, ether, dichloromethyl methyl, alpha,alpha-dichloromethyl methyl ether, methoxydichloromethane, 1,1-dichloromethyl methyl ether, alpha,alpha-dichloromethyl ether, dichloromethylmethylether
COC(Cl)Cl
114.95
238.08
∼254°C (decomposition)
MFCD03426152
UN3261
GRTGGSXWHGKRSB-UHFFFAOYSA-N
dichloro(methoxy)methane
21004
4-Bromomethyl-2(1H)-quinolinone
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.

P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c

H314-H335

DOTInformation : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.

TSCA : No

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only