missing translation for 'onlineSavingsMsg'
Learn More

4-Bromomethyl-2(1H)-quinolinone, Thermo Scientific Chemicals

Catalog No. AAB2530506
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2530506 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAB2530506 Supplier Thermo Scientific Chemicals Supplier No. B2530506

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 4876-10-2 | Molecular Formula: C2H4Cl2O | Molecular Weight: 114.95 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 4876-10-2
MDL Number MFCD03426152

Specifications

Melting Point ∼254°C (decomposition)
Molecular Formula C2H4Cl2O
Quantity 5 g
UN Number UN3261
Synonym dichloromethyl methyl ether, 1,1-dichlorodimethyl ether, dichloro methoxy methane, methane, dichloromethoxy, ether, dichloromethyl methyl, alpha,alpha-dichloromethyl methyl ether, methoxydichloromethane, 1,1-dichloromethyl methyl ether, alpha,alpha-dichloromethyl ether, dichloromethylmethylether
InChI Key GRTGGSXWHGKRSB-UHFFFAOYSA-N
SMILES COC(Cl)Cl
IUPAC Name dichloro(methoxy)methane
Molecular Weight (g/mol) 114.95
PubChem CID 21004
Formula Weight 238.08
Chemical Name or Material 4-Bromomethyl-2(1H)-quinolinone
Show More Show Less
Hazard Category H314-H335
Hazard Statement GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only