missing translation for 'onlineSavingsMsg'
Learn More

4-Bromoisoxazole, 95%

Catalog No. AC458730010
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC458730010 1 g
AC458730050 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC458730010 Supplier Thermo Scientific Chemicals Supplier No. 458730010
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 97925-43-4 | C3H2BrNO | 147.96 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 97925-43-4
Molecular Formula C3H2BrNO
Molecular Weight (g/mol) 147.96
InChI Key IRDVIGFQMLUJAM-UHFFFAOYSA-N
Synonym 4-bromoisoxazole, 4-bormoisoxazole, isoxazole, 4-bromo, isoxazole, 4-bromo-6ci,9ci, 4-bromo isoxazole, 4-bromo-isoxazole, zlchem 1329
PubChem CID 3862248
IUPAC Name 4-bromo-1,2-oxazole
SMILES C1=C(C=NO1)Br

Specifications

Melting Point 39.0°C to 41.0°C
Infrared Spectrum Authentic
Assay Percent Range 94% min. (GC)
Packaging Glass Bottle
Quantity 1 g
Formula Weight 147.96
Percent Purity 95%
Chemical Name or Material 4-Bromoisoxazole
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement Harmful if swallowed. Harmful in contact with skin. Harmful if inhaled.
Precautionary Statement GHS P Statement Wear protective gloves/protective clothing. IF SWALLOWED: rinse mouth. Do NOT induce vomiting. IF ON SKIN: Wash with plenty of soap and water. IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing. Call a POISON CENTER or doctor/physician if you feel unwell.

RUO – Research Use Only