missing translation for 'onlineSavingsMsg'
Learn More

4-Bromoisoquinoline, 98%

Catalog No. AC107090010
Change view
Click to view available options
Quantity:
1 g
10 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC107090010 1 g Glass bottle
AC107090100 10 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC107090010 Supplier Thermo Scientific Chemicals Supplier No. 107090010
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 1532-97-4 | C9H6BrN | 208.06 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1532-97-4
Molecular Formula C9H6BrN
Molecular Weight (g/mol) 208.06
MDL Number MFCD00006904
InChI Key SCRBSGZBTHKAHU-UHFFFAOYSA-N
Synonym isoquinoline, 4-bromo, 4-bromo-isoquinoline, 4-isoquinolinyl bromide, 4-bromisochinolin, 4bromoisoquinoline, 4-bromoisoquinoline, 4-bromo isoquinoline, 4-bromoisoquinoline?, pubchem6212, acmc-1bu1h
PubChem CID 73743
IUPAC Name 4-bromoisoquinoline
SMILES BrC1=C2C=CC=CC2=CN=C1

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 39.0°C to 43.0°C
Color White to Beige
Boiling Point 280.0°C to 285.0°C
Flash Point >110°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Quantity 1 g
Beilstein 20, 384
Formula Weight 208.06
Percent Purity 98%
Physical Form Crystalline Solid
Chemical Name or Material 4-Bromoisoquinoline, 98%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Causes skin irritation.
Harmful in contact with skin.
Harmful if swallowed.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 216-244-8

RUO – Research Use Only