missing translation for 'onlineSavingsMsg'
Learn More

4-Bromoisophthalic Acid 97.0+%, TCI America™

Catalog No. p-7118955
Change view
Click to view available options
Quantity:
10 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B126010G 10 g
B126025G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B126010G Supplier TCI America Supplier No. B126010G
Only null left

Chemical Identifiers

CAS 6939-93-1
Molecular Formula C8H5BrO4
Molecular Weight (g/mol) 245.028
MDL Number MFCD00002404
InChI Key MSQIEZXCNYUWHN-UHFFFAOYSA-N
Synonym 4-bromoisophthalic acid, 1,3-benzenedicarboxylic acid, 4-bromo, 4-bromoisophthalicacid, 4-bromo-1,3-benzenedicarboxylic acid, 2-bromobenzene-1,5-dicarboxylic acid, pubchem12479, chemwish ic03530, ksc354c4d, msqiezxcnyuwhn-uhfffaoysa
PubChem CID 81361
IUPAC Name 4-bromobenzene-1,3-dicarboxylic acid
SMILES C1=CC(=C(C=C1C(=O)O)C(=O)O)Br

Specifications

Color White
Quantity 10 g
Formula Weight 245.03
Percent Purity ≥97.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 4-Bromoisophthalic Acid
EINECSNumber (3)-0062
TSCA No