missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Bromoisatin 98.0+%, TCI America™
Supplier: TCI America B242325G
Specifications
| 4-Bromoisatin | |
| 271°C | |
| 25 g | |
| ITRAKBJPMLKWIW-UHFFFAOYSA-N | |
| 4-bromo-1H-indole-2,3-dione | |
| 4500012 | |
| ≥98.0% (GC,T) |
| 20780-72-7 | |
| C8H4BrNO2 | |
| 4-Bromoindole-2,3-dione, 4-Bromo-2,3-indolinedione | |
| C1=CC2=C(C(=C1)Br)C(=O)C(=O)N2 | |
| 226.029 | |
| 226.03 | |
| Crystalline Powder |
Chemical Identifiers
| 20780-72-7 | |
| 226.029 | |
| 4-Bromoindole-2,3-dione, 4-Bromo-2,3-indolinedione | |
| 4-bromo-1H-indole-2,3-dione |
| C8H4BrNO2 | |
| ITRAKBJPMLKWIW-UHFFFAOYSA-N | |
| 4500012 | |
| C1=CC2=C(C(=C1)Br)C(=O)C(=O)N2 |
Safety and Handling
TSCA : No