missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Bromoindole 97.0+%, TCI America™
$69.31 - $223.33
Chemical Identifiers
| CAS | 52488-36-5 |
|---|---|
| Molecular Formula | C8H6BrN |
| Molecular Weight (g/mol) | 196.047 |
| MDL Number | MFCD00671502 |
| InChI Key | GRJZJFUBQYULKL-UHFFFAOYSA-N |
| Synonym | 4-bromoindole, 1h-indole, 4-bromo, 4-bromo-indole, pubchem7449, acmc-1amo5, ksc269g6j, grjzjfubqyulkl-uhfffaoysa |
| PubChem CID | 676494 |
| IUPAC Name | 4-bromo-1H-indole |
| SMILES | C1=CC2=C(C=CN2)C(=C1)Br |
Chemical Identifiers
| 52488-36-5 | |
| 196.047 | |
| GRJZJFUBQYULKL-UHFFFAOYSA-N | |
| 676494 | |
| C1=CC2=C(C=CN2)C(=C1)Br |
| C8H6BrN | |
| MFCD00671502 | |
| 4-bromoindole, 1h-indole, 4-bromo, 4-bromo-indole, pubchem7449, acmc-1amo5, ksc269g6j, grjzjfubqyulkl-uhfffaoysa | |
| 4-bromo-1H-indole |
Specifications
| 52488-36-5 | |
| 96°C | |
| MFCD00671502 | |
| 4-bromoindole, 1h-indole, 4-bromo, 4-bromo-indole, pubchem7449, acmc-1amo5, ksc269g6j, grjzjfubqyulkl-uhfffaoysa | |
| C1=CC2=C(C=CN2)C(=C1)Br | |
| 196.047 | |
| 196.05 | |
| Liquid |
| Yellow | |
| C8H6BrN | |
| 1 g | |
| GRJZJFUBQYULKL-UHFFFAOYSA-N | |
| 4-bromo-1H-indole | |
| 676494 | |
| ≥97.0% (GC) | |
| 4-Bromoindole |
Safety and Handling
TSCA : No