missing translation for 'onlineSavingsMsg'
Learn More

4-Bromoindole 97.0+%, TCI America™

Catalog No. B17975G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B17975G 5 g
B17971G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B17975G Supplier TCI America Supplier No. B17975G

Chemical Identifiers

CAS 52488-36-5
Molecular Formula C8H6BrN
Molecular Weight (g/mol) 196.047
MDL Number MFCD00671502
InChI Key GRJZJFUBQYULKL-UHFFFAOYSA-N
Synonym 4-bromoindole, 1h-indole, 4-bromo, 4-bromo-indole, pubchem7449, acmc-1amo5, ksc269g6j, grjzjfubqyulkl-uhfffaoysa
PubChem CID 676494
IUPAC Name 4-bromo-1H-indole
SMILES C1=CC2=C(C=CN2)C(=C1)Br

Specifications

Color Yellow
Boiling Point 96°C
Quantity 5 g
Formula Weight 196.05
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material 4-Bromoindole