missing translation for 'onlineSavingsMsg'
Learn More

4-Bromoindole, 98%

Catalog No. AAL1941706
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1941706 5 g
AAL1941703 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1941706 Supplier Thermo Scientific Chemicals Supplier No. L1941706
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 52488-36-5 | C8H6BrN | 196.047 g/mol

4-Bromoindole is a potential inhibitor of GSK-3. It is also used as pharmaceutical intermediates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Bromoindole is a potential inhibitor of GSK-3. It is also used as pharmaceutical intermediates.

Solubility
Not miscible in water.

Notes
Air & Light Sensitive. Store at 4°C. Store in the dark. Store under dry inert gas. Protect from heat. Incompatible with heat. light, air.

Chemical Identifiers

CAS 52488-36-5
Molecular Formula C8H6BrN
Molecular Weight (g/mol) 196.047
MDL Number MFCD00671502
InChI Key GRJZJFUBQYULKL-UHFFFAOYSA-N
Synonym 4-bromoindole, 1h-indole, 4-bromo, 4-bromo-indole, pubchem7449, acmc-1amo5, ksc269g6j, grjzjfubqyulkl-uhfffaoysa
PubChem CID 676494
IUPAC Name 4-bromo-1H-indole
SMILES C1=CC2=C(C=CN2)C(=C1)Br

Specifications

Melting Point 17°C
Density 1.563
Boiling Point 88°C to 90°C (0.1 mmHg)
Flash Point >110°C (230°F)
Refractive Index 1.6545
Quantity 5 g
Beilstein 114891
Sensitivity Air and light sensitive
Solubility Information Not miscible in water.
Formula Weight 196.05
Percent Purity 98%
Chemical Name or Material 4-Bromoindole
Show More Show Less
Hazard Category H302+H312+H332-H315-H319-H335
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
DOTInformation Hazard Class: 6.1; Packaging Group: III
EINECSNumber 000-000-0
TSCA No
Recommended Storage Keep cold; Store under Nitrogen

RUO – Research Use Only