missing translation for 'onlineSavingsMsg'
Learn More

4-Bromoindole-3-carboxaldehyde 96.0+%, TCI America™

Catalog No. B27731G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B27731G 1 g
B27735G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B27731G Supplier TCI America Supplier No. B27731G

Chemical Identifiers

CAS 98600-34-1
Molecular Formula C9H6BrNO
Molecular Weight (g/mol) 224.057
InChI Key IPAFHZDRYAWZOB-UHFFFAOYSA-N
Synonym 4-bromoindole-3-carboxaldehyde, 4-bromoindole-3-carboxyaldehyde, 4-bromo-3-formylindole, 4-bromo-1h-indole-3-carboxaldehyde, 4-bromoindole-3-carbaldehyde, 4-bromo-3-formyl-1h-indole, 1h-indole-3-carboxaldehyde, 4-bromo, 4-bromo-3-indolecarbaldehyde, pubchem7682, acmc-209saq
PubChem CID 2763178
IUPAC Name 4-bromo-1H-indole-3-carbaldehyde
SMILES C1=CC2=C(C(=C1)Br)C(=CN2)C=O

Specifications

Melting Point 179°C
Color Yellow
Quantity 1 g
Formula Weight 224.06
Percent Purity ≥96.0% (GC,N)
Physical Form Crystalline Powder
Chemical Name or Material 4-Bromoindole-3-carboxaldehyde