missing translation for 'onlineSavingsMsg'
Learn More

4-Bromoindole-3-carboxaldehyde, 97%, Thermo Scientific Chemicals

Catalog No. p-7057366
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6696403 1 g
AAH6696406 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH6696403 Supplier Thermo Scientific Chemicals Supplier No. H6696403
Only null left

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 98600-34-1
Molecular Formula C9H6BrNO
Molecular Weight (g/mol) 224.057
MDL Number MFCD05864695
InChI Key IPAFHZDRYAWZOB-UHFFFAOYSA-N
Synonym 4-bromoindole-3-carboxaldehyde, 4-bromoindole-3-carboxyaldehyde, 4-bromo-3-formylindole, 4-bromo-1h-indole-3-carboxaldehyde, 4-bromoindole-3-carbaldehyde, 4-bromo-3-formyl-1h-indole, 1h-indole-3-carboxaldehyde, 4-bromo, 4-bromo-3-indolecarbaldehyde, pubchem7682, acmc-209saq
PubChem CID 2763178
IUPAC Name 4-bromo-1H-indole-3-carbaldehyde
SMILES C1=CC2=C(C(=C1)Br)C(=CN2)C=O

Specifications

Quantity 1 g
Beilstein 5733778
Formula Weight 224.06
Percent Purity 97%
Physical Form Solid
Chemical Name or Material 4-Bromoindole-3-carboxaldehyde
Hazard Category H302-H315-H319-H335
Hazard Statement GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only