missing translation for 'onlineSavingsMsg'
Learn More

4-Bromoindazole 98.0+%, TCI America™

Catalog No. B46315G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B46315G 5 g
B46311G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B46315G Supplier TCI America Supplier No. B46315G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 186407-74-9
Molecular Formula C7H5BrN2
Molecular Weight (g/mol) 197.035
MDL Number MFCD05664001
InChI Key KJIODOACRIRBPB-UHFFFAOYSA-N
Synonym 4-bromoindazole, 1h-indazole, 4-bromo, 4-bromo 1h indazole, 4-bromo-indazole, bromo-1h-indazole, pubchem7834, 4-bromo-2h-indazole, acmc-1bzds, 4-bromanyl-1h-indazole
PubChem CID 22352548
IUPAC Name 4-bromo-1H-indazole
SMILES C1=CC2=C(C=NN2)C(=C1)Br

Specifications

Melting Point 167°C
Color Yellow
Quantity 5 g
Formula Weight 197.04
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4-Bromoindazole