missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Bromodibenzofuran 98.0+%, TCI America™
$145.40 - $490.25
Chemical Identifiers
| CAS | 89827-45-2 |
|---|---|
| Molecular Formula | C12H7BrO |
| Molecular Weight (g/mol) | 247.091 |
| MDL Number | MFCD00185685 |
| InChI Key | DYTYBRPMNQQFFL-UHFFFAOYSA-N |
| PubChem CID | 458256 |
| IUPAC Name | 4-bromodibenzofuran |
| SMILES | C1=CC=C2C(=C1)C3=C(O2)C(=CC=C3)Br |
Chemical Identifiers
| 89827-45-2 | |
| 247.091 | |
| DYTYBRPMNQQFFL-UHFFFAOYSA-N | |
| 4-bromodibenzofuran |
| C12H7BrO | |
| MFCD00185685 | |
| 458256 | |
| C1=CC=C2C(=C1)C3=C(O2)C(=CC=C3)Br |
Specifications
| 89827-45-2 | |
| White-Yellow | |
| MFCD00185685 | |
| DYTYBRPMNQQFFL-UHFFFAOYSA-N | |
| 4-bromodibenzofuran | |
| 458256 | |
| ≥98.0% (GC) | |
| 4-Bromodibenzofuran |
| 70°C | |
| C12H7BrO | |
| 1 g | |
| C1=CC=C2C(=C1)C3=C(O2)C(=CC=C3)Br | |
| 247.091 | |
| 247.09 | |
| Crystalline Powder |
Safety and Handling
TSCA : No