missing translation for 'onlineSavingsMsg'
Learn More

4-Bromocinnamic Acid 98.0+%, TCI America™

Catalog No. B16905G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B16905G 5 g
B169025G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B16905G Supplier TCI America Supplier No. B16905G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 1200-07-3
Molecular Formula C9H7BrO2
Molecular Weight (g/mol) 227.06
MDL Number MFCD00004394
InChI Key CPDDDTNAMBSPRN-ZZXKWVIFSA-N
Synonym 4-bromocinnamic acid, p-bromocinnamic acid, 3-4-bromophenyl acrylic acid, bromocinnamic acid,4, 2e-3-4-bromophenyl prop-2-enoic acid, trans-4-bromocinnamic acid, e-3-4-bromophenyl acrylic acid, 2-propenoic acid, 3-4-bromophenyl-, 2e, 4-bromocinnamic acid, predominantly trans, rarechem bk hd c006
PubChem CID 737158
IUPAC Name (2E)-3-(4-bromophenyl)prop-2-enoic acid
SMILES OC(=O)\C=C\C1=CC=C(Br)C=C1

Specifications

Quantity 5 g
Formula Weight 227.06
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 4-Bromocinnamic Acid