missing translation for 'onlineSavingsMsg'
Learn More

4-Bromocinnamic acid, predominantly trans, 97+%

Catalog No. AAA1150322
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1150322 100 g
AAA1150306 5 g
AAA1150314 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1150322 Supplier Thermo Scientific Chemicals Supplier No. A1150322
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 1200-07-3 | C9H7BrO2 | 227.06 g/mol

4-Bromocinnamic acid has been used in the preparation of: (E)-β-bromo-4-bromostyrene, 2-amino-7-(piperidin-4-yl)isoquinoline, brominated dansyl derivative (4-bromophenyl)-4-(5-(dimethylamino)naphthalene-1-sulfonamido)butanoic acid.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Bromocinnamic acid has been used in the preparation of: (E)-β-bromo-4-bromostyrene, 2-amino-7-(piperidin-4-yl)isoquinoline, brominated dansyl derivative (4-bromophenyl)-4-(5-(dimethylamino)naphthalene-1-sulfonamido)butanoic acid.

Solubility
Soluble in alcohol, ethyl acetate, DMSO. Insoluble in water.

Notes
Store at -20°C. Store in a cool, dry, well-ventilated area away from incompatible substances such as oxidizing agents.

Chemical Identifiers

CAS 1200-07-3
Molecular Formula C9H7BrO2
Molecular Weight (g/mol) 227.06
MDL Number MFCD00004394
InChI Key CPDDDTNAMBSPRN-ZZXKWVIFSA-N
Synonym 4-bromocinnamic acid, p-bromocinnamic acid, 3-4-bromophenyl acrylic acid, bromocinnamic acid,4, 2e-3-4-bromophenyl prop-2-enoic acid, trans-4-bromocinnamic acid, e-3-4-bromophenyl acrylic acid, 2-propenoic acid, 3-4-bromophenyl-, 2e, 4-bromocinnamic acid, predominantly trans, rarechem bk hd c006
PubChem CID 737158
IUPAC Name (E)-3-(4-bromophenyl)prop-2-enoic acid
SMILES OC(=O)\C=C\C1=CC=C(Br)C=C1

Specifications

Melting Point 260°C to 264°C
Odor Odorless
Quantity 100 g
Beilstein 3538823
Solubility Information Soluble in alcohol,ethyl acetate,DMSO. Insoluble in water.
Formula Weight 227.06
Percent Purity ≥98%
Chemical Name or Material 4-Bromocinnamic acid, predominantly trans
Hazard Category H302-H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
EINECSNumber 214-850-7
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only