missing translation for 'onlineSavingsMsg'
Learn More

4-Bromobutyryl chloride, 95%

Catalog No. AC303440050
Change view
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC303440050 5 g Glass bottle
AC303440250 25 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC303440050 Supplier Thermo Scientific Chemicals Supplier No. 303440050
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 927-58-2 | C4H6BrClO | 185.46 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 927-58-2
Molecular Formula C4H6BrClO
Molecular Weight (g/mol) 185.46
MDL Number MFCD00000753
InChI Key LRTRXDSAJLSRTG-UHFFFAOYSA-N
Synonym 4-bromobutyryl chloride, butanoyl chloride, 4-bromo, g-bromobutyroyl chloride, butyryl chloride, 4-bromo, 4-bromobutyrylchloride, bromobutryl chloride, bromobutyryl chloride, acmc-20ammt, 4-bromobutanoylchloride, 4-bromobutyroylchloride
PubChem CID 70225
IUPAC Name 4-bromobutanoyl chloride
SMILES C(CC(=O)Cl)CBr

Specifications

CAS Min % 94.0
CAS Max % 100.0
Color Colorless to Yellow or Pink
Density 1.6020g/mL
Boiling Point 101°C (37.0 mmHg)
Flash Point 91°C
Infrared Spectrum Authentic
Assay Percent Range 94% min. (GC)
Packaging Glass bottle
Refractive Index 1.4910 to 1.494
Linear Formula Br(CH2)3COCl
Quantity 5 g
Specific Gravity 1.602
Formula Weight 185.46
Percent Purity 95%
Physical Form Liquid
Chemical Name or Material 4-Bromobutyryl chloride
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Causes severe skin burns and eye damage.
Reacts violently with water.
Precautionary Statement GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsi
EINECSNumber 213-154-

RUO – Research Use Only