missing translation for 'onlineSavingsMsg'
Learn More

4-Bromobutylboronic acid (1S,2S,3R,5S)-(+)-2,3-pinanediol ester, 95%, Thermo Scientific Chemicals

$204.78 - $204.78

Chemical Identifiers

CAS 165881-36-7
Molecular Formula C14H24BBrO2
Molecular Weight (g/mol) 315.058
MDL Number MFCD09037500
InChI Key DQGAIMVYOVPBSD-FMSGJZPZSA-N
Synonym 4-bromobutylboronic acid 1s,2s,3r,5s-+-2,3-pinanediol ester, 4-bromobutylboronate ester, 4-bromobutaneboronic acid 1s,2s,3r,5s-+-2,3-pinanediol ester, 1s,2s,3r,5s-+-2,3-pinanediol 4-bromobutylboronate ester, 1s,2s,6r,8s-4-4-bromobutyl-1,8-dihydrogenio-2,9,9-trimethyl-3,5-dioxa-4-boratricyclo 6.1.1.02,6 decane, 1s,2s,6r,8s-4-4-bromobutyl-2,9,9-trimethyl-3,5-dioxa-4-boratricyclo 6.1.1.0?,? decane, 1s,2s,6r,8s-4-4-bromobutyl-2,9,9-trimethyl-3,5-dioxa-4-boratricyclo 6.1.1.0^ 2,6 decane, 1s,2s,6r,8s-4-4-bromobutyl-2,9,9-trimethyl-3,5-dioxa-4-boratricyclo 6.1.1.0_,? decane, 3as,4s,6s,7ar-2-4-bromobutyl-3a,5,5-trimethylhexahydro-2h-4,6-methano-1,3,2-benzodioxaborole, 3as,4s,6s,7ar-2-4-bromobutyl-3a,5,5-trimethylhexahydro-4,6-methanobenzo d 1,3,2 dioxaborole
PubChem CID 44784918
SMILES B1(OC2CC3CC(C3(C)C)C2(O1)C)CCCCBr
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH2709703
View Documents
Thermo Scientific Chemicals
H2709703
1 g
Each for $204.78
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

165881-36-7
315.058
DQGAIMVYOVPBSD-FMSGJZPZSA-N
44784918
C14H24BBrO2
MFCD09037500
4-bromobutylboronic acid 1s,2s,3r,5s-+-2,3-pinanediol ester, 4-bromobutylboronate ester, 4-bromobutaneboronic acid 1s,2s,3r,5s-+-2,3-pinanediol ester, 1s,2s,3r,5s-+-2,3-pinanediol 4-bromobutylboronate ester, 1s,2s,6r,8s-4-4-bromobutyl-1,8-dihydrogenio-2,9,9-trimethyl-3,5-dioxa-4-boratricyclo 6.1.1.02,6 decane, 1s,2s,6r,8s-4-4-bromobutyl-2,9,9-trimethyl-3,5-dioxa-4-boratricyclo 6.1.1.0?,? decane, 1s,2s,6r,8s-4-4-bromobutyl-2,9,9-trimethyl-3,5-dioxa-4-boratricyclo 6.1.1.0^ 2,6 decane, 1s,2s,6r,8s-4-4-bromobutyl-2,9,9-trimethyl-3,5-dioxa-4-boratricyclo 6.1.1.0_,? decane, 3as,4s,6s,7ar-2-4-bromobutyl-3a,5,5-trimethylhexahydro-2h-4,6-methano-1,3,2-benzodioxaborole, 3as,4s,6s,7ar-2-4-bromobutyl-3a,5,5-trimethylhexahydro-4,6-methanobenzo d 1,3,2 dioxaborole
B1(OC2CC3CC(C3(C)C)C2(O1)C)CCCCBr

Specifications

165881-36-7
1.496
1 g
DQGAIMVYOVPBSD-FMSGJZPZSA-N
B1(OC2CC3CC(C3(C)C)C2(O1)C)CCCCBr
44784918
95%
C14H24BBrO2
MFCD09037500
4-bromobutylboronic acid 1s,2s,3r,5s-+-2,3-pinanediol ester, 4-bromobutylboronate ester, 4-bromobutaneboronic acid 1s,2s,3r,5s-+-2,3-pinanediol ester, 1s,2s,3r,5s-+-2,3-pinanediol 4-bromobutylboronate ester, 1s,2s,6r,8s-4-4-bromobutyl-1,8-dihydrogenio-2,9,9-trimethyl-3,5-dioxa-4-boratricyclo 6.1.1.02,6 decane, 1s,2s,6r,8s-4-4-bromobutyl-2,9,9-trimethyl-3,5-dioxa-4-boratricyclo 6.1.1.0?,? decane, 1s,2s,6r,8s-4-4-bromobutyl-2,9,9-trimethyl-3,5-dioxa-4-boratricyclo 6.1.1.0^ 2,6 decane, 1s,2s,6r,8s-4-4-bromobutyl-2,9,9-trimethyl-3,5-dioxa-4-boratricyclo 6.1.1.0_,? decane, 3as,4s,6s,7ar-2-4-bromobutyl-3a,5,5-trimethylhexahydro-2h-4,6-methano-1,3,2-benzodioxaborole, 3as,4s,6s,7ar-2-4-bromobutyl-3a,5,5-trimethylhexahydro-4,6-methanobenzo d 1,3,2 dioxaborole
+18° (c=1 in Chloroform)
315.058
315.06
4-Bromobutylboronic acid (1S,2S,3R,5S)-(+)-2,3-pinanediol ester
Videos
Safety and Handling

Safety and Handling

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

RUO – Research Use Only