missing translation for 'onlineSavingsMsg'
Learn More

4-Bromobutylboronic acid (1S,2S,3R,5S)-(+)-2,3-pinanediol ester, 95%, Thermo Scientific Chemicals

Catalog No. AAH2709703
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH2709703 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH2709703 Supplier Thermo Scientific Chemicals Supplier No. H2709703
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 165881-36-7 | Molecular Formula: C14H24BBrO2 | Molecular Weight: 315.058 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 165881-36-7
Molecular Formula C14H24BBrO2
Molecular Weight (g/mol) 315.058
MDL Number MFCD09037500
InChI Key DQGAIMVYOVPBSD-FMSGJZPZSA-N
Synonym 4-bromobutylboronic acid 1s,2s,3r,5s-+-2,3-pinanediol ester, 4-bromobutylboronate ester, 4-bromobutaneboronic acid 1s,2s,3r,5s-+-2,3-pinanediol ester, 1s,2s,3r,5s-+-2,3-pinanediol 4-bromobutylboronate ester, 1s,2s,6r,8s-4-4-bromobutyl-1,8-dihydrogenio-2,9,9-trimethyl-3,5-dioxa-4-boratricyclo 6.1.1.02,6 decane, 1s,2s,6r,8s-4-4-bromobutyl-2,9,9-trimethyl-3,5-dioxa-4-boratricyclo 6.1.1.0?,? decane, 1s,2s,6r,8s-4-4-bromobutyl-2,9,9-trimethyl-3,5-dioxa-4-boratricyclo 6.1.1.0^ 2,6 decane, 1s,2s,6r,8s-4-4-bromobutyl-2,9,9-trimethyl-3,5-dioxa-4-boratricyclo 6.1.1.0_,? decane, 3as,4s,6s,7ar-2-4-bromobutyl-3a,5,5-trimethylhexahydro-2h-4,6-methano-1,3,2-benzodioxaborole, 3as,4s,6s,7ar-2-4-bromobutyl-3a,5,5-trimethylhexahydro-4,6-methanobenzo d 1,3,2 dioxaborole
PubChem CID 44784918
SMILES B1(OC2CC3CC(C3(C)C)C2(O1)C)CCCCBr

Specifications

Refractive Index 1.496
Quantity 1 g
Optical Rotation +18° (c=1 in Chloroform)
Formula Weight 315.06
Percent Purity 95%
Chemical Name or Material 4-Bromobutylboronic acid (1S,2S,3R,5S)-(+)-2,3-pinanediol ester
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only