missing translation for 'onlineSavingsMsg'
Learn More

4-Bromobenzylamine, 97+%

CAS: 3959-07-7 | C7H8BrN | 186.05 g/mol

$384.31 - $384.31

Chemical Identifiers

CAS 3959-07-7
Molecular Formula C7H8BrN
Molecular Weight (g/mol) 186.05
MDL Number MFCD00047931
InChI Key XRNVSPDQTPVECU-UHFFFAOYSA-N
SMILES NCC1=CC=C(Br)C=C1
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC350530050
SDS View Documents
Thermo Scientific Chemicals
350530050
5 g Glass bottle N/A
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

3959-07-7
186.05
XRNVSPDQTPVECU-UHFFFAOYSA-N
C7H8BrN
MFCD00047931
NCC1=CC=C(Br)C=C1

Specifications

3959-07-7
100.0
Colorless
Authentic
Glass bottle
1.5855 to 1.5875
MFCD00047931
12, III, 2350
XRNVSPDQTPVECU-UHFFFAOYSA-N
(2-bromophenyl)methanamine
334072
97+%
4-Bromobenzylamine
98.5
25.0°C
>110°C
97% min. (GC)
C7H8BrN
BrC6H4CH2NH2
5 g
2-bromobenzylamine, 2-bromophenyl methanamine, o-bromobenzylamine, 2-bromophenyl methylamine, 1-2-bromophenyl methanamine, 2-brobenzylamine, 2-bromo-benzylamine, pubchem20398, acmc-1ct06, benzenemethanamine, 2-bromo
NCC1=CC=C(Br)C=C1
186.05
186.05
Liquid
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Causes severe skin burns and eye damage.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with wat

GHS Signal Word: Danger

EINECSNumber : 223-559-4

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only