missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4'-Bromobenzo-15-crown 5-Ether 93.0+%, TCI America™
$467.87 - $1612.41
Chemical Identifiers
| CAS | 60835-72-5 |
|---|---|
| Molecular Formula | C14H19BrO5 |
| Molecular Weight (g/mol) | 347.205 |
| MDL Number | MFCD00143270 |
| InChI Key | GPKJNSIFVWMEEI-UHFFFAOYSA-N |
| Synonym | m-bromobenzo-15-crown-5, 4'-bromobenzo-15-crown 5-ether, 4'-bromobenzo-15-crown-5, 1,4,7,10,13-benzopentaoxacyclopentadecin, 2,3,5,6,8,9,11,12-octahydro-15-bromo, 15-bromo-2,3,5,6,8,9,11,12-octahydro-1,4,7,10,13-benzopentaoxacyclopentadecine, 2,3,5,6,8,9,11,12-octahydro-15-bromo-1,4,7,10,13-benzopentaoxacyclopentadecin, 1,2-oxybis ethyleneoxyethyleneoxy-4-bromobenzene, 4 inverted exclamation marka-bromobenzo-15-crown-5, benzo-15-crown-5-4 inverted exclamation marka-yl bromide, 2,3-4-bromobenzo-1,4,7,10,13-pentaoxacyclopentadec-2-ene |
| PubChem CID | 2724799 |
| IUPAC Name | 17-bromo-2,5,8,11,14-pentaoxabicyclo[13.4.0]nonadeca-1(15),16,18-triene |
| SMILES | C1COCCOC2=C(C=C(C=C2)Br)OCCOCCO1 |
Chemical Identifiers
| 60835-72-5 | |
| 347.205 | |
| GPKJNSIFVWMEEI-UHFFFAOYSA-N | |
| 2724799 | |
| C1COCCOC2=C(C=C(C=C2)Br)OCCOCCO1 |
| C14H19BrO5 | |
| MFCD00143270 | |
| m-bromobenzo-15-crown-5, 4'-bromobenzo-15-crown 5-ether, 4'-bromobenzo-15-crown-5, 1,4,7,10,13-benzopentaoxacyclopentadecin, 2,3,5,6,8,9,11,12-octahydro-15-bromo, 15-bromo-2,3,5,6,8,9,11,12-octahydro-1,4,7,10,13-benzopentaoxacyclopentadecine, 2,3,5,6,8,9,11,12-octahydro-15-bromo-1,4,7,10,13-benzopentaoxacyclopentadecin, 1,2-oxybis ethyleneoxyethyleneoxy-4-bromobenzene, 4 inverted exclamation marka-bromobenzo-15-crown-5, benzo-15-crown-5-4 inverted exclamation marka-yl bromide, 2,3-4-bromobenzo-1,4,7,10,13-pentaoxacyclopentadec-2-ene | |
| 17-bromo-2,5,8,11,14-pentaoxabicyclo[13.4.0]nonadeca-1(15),16,18-triene |
Specifications
| 60835-72-5 | |
| C14H19BrO5 | |
| 1 g | |
| GPKJNSIFVWMEEI-UHFFFAOYSA-N | |
| 17-bromo-2,5,8,11,14-pentaoxabicyclo[13.4.0]nonadeca-1(15),16,18-triene | |
| 2724799 | |
| ≥93.0% (GC) | |
| 4′-Bromobenzo-15-crown 5-Ether |
| 70°C | |
| MFCD00143270 | |
| m-bromobenzo-15-crown-5, 4'-bromobenzo-15-crown 5-ether, 4'-bromobenzo-15-crown-5, 1,4,7,10,13-benzopentaoxacyclopentadecin, 2,3,5,6,8,9,11,12-octahydro-15-bromo, 15-bromo-2,3,5,6,8,9,11,12-octahydro-1,4,7,10,13-benzopentaoxacyclopentadecine, 2,3,5,6,8,9,11,12-octahydro-15-bromo-1,4,7,10,13-benzopentaoxacyclopentadecin, 1,2-oxybis ethyleneoxyethyleneoxy-4-bromobenzene, 4 inverted exclamation marka-bromobenzo-15-crown-5, benzo-15-crown-5-4 inverted exclamation marka-yl bromide, 2,3-4-bromobenzo-1,4,7,10,13-pentaoxacyclopentadec-2-ene | |
| C1COCCOC2=C(C=C(C=C2)Br)OCCOCCO1 | |
| 347.205 | |
| 347.21 | |
| Crystalline Powder |
Safety and Handling
RTECSNumber : DI5450000
TSCA : No