missing translation for 'onlineSavingsMsg'
Learn More

4'-Bromobenzo-15-crown 5-Ether 93.0+%, TCI America™

Catalog No. B21895G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B21895G 5 g
B21891G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B21895G Supplier TCI America Supplier No. B21895G
Only null left

Chemical Identifiers

CAS 60835-72-5
Molecular Formula C14H19BrO5
Molecular Weight (g/mol) 347.205
MDL Number MFCD00143270
InChI Key GPKJNSIFVWMEEI-UHFFFAOYSA-N
Synonym m-bromobenzo-15-crown-5, 4'-bromobenzo-15-crown 5-ether, 4'-bromobenzo-15-crown-5, 1,4,7,10,13-benzopentaoxacyclopentadecin, 2,3,5,6,8,9,11,12-octahydro-15-bromo, 15-bromo-2,3,5,6,8,9,11,12-octahydro-1,4,7,10,13-benzopentaoxacyclopentadecine, 2,3,5,6,8,9,11,12-octahydro-15-bromo-1,4,7,10,13-benzopentaoxacyclopentadecin, 1,2-oxybis ethyleneoxyethyleneoxy-4-bromobenzene, 4 inverted exclamation marka-bromobenzo-15-crown-5, benzo-15-crown-5-4 inverted exclamation marka-yl bromide, 2,3-4-bromobenzo-1,4,7,10,13-pentaoxacyclopentadec-2-ene
PubChem CID 2724799
IUPAC Name 17-bromo-2,5,8,11,14-pentaoxabicyclo[13.4.0]nonadeca-1(15),16,18-triene
SMILES C1COCCOC2=C(C=C(C=C2)Br)OCCOCCO1

Specifications

Melting Point 70°C
Quantity 5 g
Formula Weight 347.21
Percent Purity ≥93.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4′-Bromobenzo-15-crown 5-Ether
RTECSNumber DI5450000
TSCA No