missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4'-Bromobenzo-15-crown 5-Ether 93.0+%, TCI America™
Specifications
Specifications
| Melting Point | 70°C |
| Quantity | 1 g |
| Formula Weight | 347.21 |
| Percent Purity | ≥93.0% (GC) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | 4′-Bromobenzo-15-crown 5-Ether |
Safety and Handling
| RTECSNumber | DI5450000 |
| TSCA | No |