missing translation for 'onlineSavingsMsg'
Learn More

4-Bromobenzenesulfonyl chloride, 98%

Catalog No. p-73075
Change view
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC148760250 25 g Glass bottle
AC148761000 100 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC148760250 Supplier Thermo Scientific Chemicals Supplier No. 148760250
Only null left

CAS: 98-58-8 | C6H4BrClO2S | 255.51 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 98-58-8
Molecular Formula C6H4BrClO2S
Molecular Weight (g/mol) 255.51
MDL Number MFCD00007437
InChI Key KMMHZIBWCXYAAH-UHFFFAOYSA-N
Synonym 4-bromobenzene-1-sulfonyl chloride, p-bromobenzenesulfonyl chloride, 4-bromo-benzenesulfonyl chloride, p-bromophenylsulfonyl chloride, benzenesulfonyl chloride, 4-bromo, 4-bromobenzenesulphonyl chloride, 4-bromobenzenesulfonylchloride, unii-c6347t8fp1, 4-bromobenzene sulfonyl chloride, 4-bromophenyl chlorosulfone
PubChem CID 7396
IUPAC Name 4-bromobenzenesulfonyl chloride
SMILES C1=CC(=CC=C1S(=O)(=O)Cl)Br

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 74°C to 77°C
Color White to Beige
Boiling Point 153°C (15.0 mmHg)
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (Ex CL) (Argentometry)
Packaging Glass bottle
Linear Formula BrC6H4SO2Cl
Quantity 25 g
Beilstein 11, 57
Merck Index 15, 1414
Solubility Information Solubility in water: decomposes
Formula Weight 255.51
Percent Purity 98%
Physical Form Crystalline Powder or Crystals
Chemical Name or Material 4-Bromobenzenesulfonyl chloride, 98%
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement:
Causes serious eye damage.
Causes severe skin burns and eye damage.
Precautionary Statement GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 202-683-2
TSCA TSCA

RUO – Research Use Only