missing translation for 'onlineSavingsMsg'
Learn More

4'-Bromoacetanilide, 98%

Catalog No. AAL0292430
Change view
Click to view available options
Quantity:
50 g
250 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0292430 250 g
AAL0292418 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0292430 Supplier Thermo Scientific Chemicals Supplier No. L0292430
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 103-88-8 | C8H8BrNO | 214.06 g/mol

4'-Bromoacetanilide is used as an internal standard for phenylurea and the respective metabolic products in oysters. It has also been used as a reagent in the production of a new ligand to form a coordination compex between gadolinium(III) and T=titanium(IV) oxide. It was used in the synthesis of new ligand for anchoring Gd(III) chelates onto TiO2 surface.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4′-Bromoacetanilide is used as an internal standard for phenylurea and the respective metabolic products in oysters. It has also been used as a reagent in the production of a new ligand to form a coordination compex between gadolinium(III) and T=titanium(IV) oxide. It was used in the synthesis of new ligand for anchoring Gd(III) chelates onto TiO2 surface.

Solubility
Moderately soluble in alcohol. Insoluble in water.

Notes
Incompatible materials are Strong oxidizing agents. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store at room temperature.

Chemical Identifiers

CAS 103-88-8
Molecular Formula C8H8BrNO
Molecular Weight (g/mol) 214.06
MDL Number MFCD00000092
InChI Key MSLICLMCQYQNPK-UHFFFAOYSA-N
Synonym 4'-bromoacetanilide, n-4-bromophenyl acetamide, p-bromoacetanilide, 4-bromoacetanilide, bromoanilide, acetamide, n-4-bromophenyl, antisepsin, bromoantifebrin, asepsin, p-bromo-n-acetanilide
PubChem CID 7683
IUPAC Name N-(4-bromophenyl)acetamide
SMILES CC(=O)NC1=CC=C(Br)C=C1

Specifications

Melting Point 166°C to 171°C
Odor Odorless
Quantity 250 g
Beilstein 2208091
Merck Index 14,1399
Solubility Information Moderately soluble in alcohol. Insoluble in water.
Formula Weight 214.06
Percent Purity 98%
Chemical Name or Material 4'-Bromoacetanilide
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 203-154-9
RTECSNumber AD9625000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only