missing translation for 'onlineSavingsMsg'
Learn More

4-Bromo-N-methylphthalimide 98.0+%, TCI America™

Catalog No. B34435G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B34435G 5 g
B344325G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B34435G Supplier TCI America Supplier No. B34435G
Only null left

Chemical Identifiers

CAS 90224-73-0
Molecular Formula C9H6BrNO2
Molecular Weight (g/mol) 240.056
InChI Key BEIQHTQYTDPHLX-UHFFFAOYSA-N
PubChem CID 790105
IUPAC Name 5-bromo-2-methylisoindole-1,3-dione
SMILES CN1C(=O)C2=C(C1=O)C=C(C=C2)Br

Specifications

Melting Point 150°C
Color White-Yellow
Quantity 5 g
Formula Weight 240.06
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4-Bromo-N-methylphthalimide