missing translation for 'onlineSavingsMsg'
Learn More

4-Bromo-5-chlorothiophene-2-sulfonyl chloride, ≥95%, Thermo Scientific™

Catalog No. p-2701612
Change view
Click to view available options
Quantity:
250mg
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
SEW02652CB 250mg
SEW02652DA 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. SEW02652CB Supplier Thermo Scientific Chemicals Supplier No. SEW02652CB

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 166964-35-8
Molecular Formula C4HBrCl2O2S2
Molecular Weight (g/mol) 295.974
MDL Number MFCD00052094
InChI Key PABOKJJDABSQCK-UHFFFAOYSA-N
Synonym 3-bromo-2-chlorothiophene-5-sulfonyl chloride, 4-bromo-5-chlorothiophene-2-sulphonyl chloride, 4-bromo-5-chloro-2-thiophenesulfonyl chloride, 4-bromo-5-chlorothiophene-2-sulphonylchloride, 3-bromo-2-chlorothiophene-5-sulphonyl chloride, 2-thiophenesulfonylchloride, 4-bromo-5-chloro, pubchem5475, 4-bromo-5-chloro-thiophene-2-sulfonyl chloride, 4-bromo-5-chlorothiophene-2-sulfonyl, 4-bromanyl-5-chloranyl-thiophene-2-sulfonyl chloride
PubChem CID 2797220
IUPAC Name 4-bromo-5-chlorothiophene-2-sulfonyl chloride
SMILES C1=C(SC(=C1Br)Cl)S(=O)(=O)Cl

Specifications

CAS Min % 95.0
Assay Percent Range 95%
Packaging Amber Glass Bottle
Quantity 250mg
Formula Weight 295.99
Percent Purity 95%
Chemical Name or Material 4-Bromo-5-chlorothiophene-2-sulfonyl chloride
Hazard Statement GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.