missing translation for 'onlineSavingsMsg'
Learn More

4-Bromo-4'-[di(p-tolyl)amino]stilbene 97.0+%, TCI America™

Catalog No. b49061g
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B49061G 1 g
B4906200MG 200 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B49061G Supplier TCI America Supplier No. B49061G
Only null left

Chemical Identifiers

CAS 101186-77-0
Molecular Formula C28H24BrN
Molecular Weight (g/mol) 454.411
MDL Number MFCD28975103
InChI Key UDHCDVGKGPSNLA-UHFFFAOYSA-N
Synonym 4-(4-Bromostyryl)-N,N-di(p-tolyl)aniline, 4-(4-Bromostyryl)-4′C,4′C′C-dimethyltriphenylamine
PubChem CID 54347443
IUPAC Name N-[4-[2-(4-bromophenyl)ethenyl]phenyl]-4-methyl-N-(4-methylphenyl)aniline
SMILES CC1=CC=C(C=C1)N(C2=CC=C(C=C2)C)C3=CC=C(C=C3)C=CC4=CC=C(C=C4)Br

Specifications

Melting Point 157°C
Color Yellow
Quantity 1 g
Formula Weight 454.41
Percent Purity ≥97.0% (HPLC,N)
Physical Form Crystalline Powder
Chemical Name or Material 4-Bromo-4′-[di(p-tolyl)amino]stilbene