missing translation for 'onlineSavingsMsg'
Learn More

4'-Bromo-4-chlorobutyrophenone, 98%

Catalog No. AAB2414622
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2414622 100 g
AAB2414614 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB2414622 Supplier Thermo Scientific Chemicals Supplier No. B2414622
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 4559-96-0 | C10H10BrClO | 261.54 g/mol

4'-Bromo-4-chlorobutyrophenone is used as a pharmaceutical & syntheses material intermediates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4′-Bromo-4-chlorobutyrophenone is used as a pharmaceutical & syntheses material intermediates.

Solubility
Insoluble in water.

Notes
Storage temperature is 2 - 4°Cand Keep container tightly closed in a dryand well-ventilated place.

Chemical Identifiers

CAS 4559-96-0
Molecular Formula C10H10BrClO
Molecular Weight (g/mol) 261.54
MDL Number MFCD00001006
InChI Key WJKPUMBLABGUCQ-UHFFFAOYSA-N
Synonym 1-4-bromophenyl-4-chlorobutan-1-one, 4'-bromo-4-chlorobutyrophenone, 1-4-bromophenyl-4-chloro-1-butanone, 1-butanone, 1-4-bromophenyl-4-chloro, acmc-20amn1, 4-chloro-p-bromobutyrophenone, 4-chloro-4'-bromobutyrophenone, 4-bromo-4-chlorobutyrophenone, 4/'-bromo-4-chlorobutyrophenone, 4'-bromo-gamma-chlorobutyrophenone
PubChem CID 78317
IUPAC Name 1-(4-bromophenyl)-4-chlorobutan-1-one
SMILES ClCCCC(=O)C1=CC=C(Br)C=C1

Specifications

Melting Point 35°C to 38°C
Flash Point >110°C (230°F)
Quantity 100 g
Solubility Information Insoluble in water.
Formula Weight 261.55
Percent Purity 98%
Chemical Name or Material 4'-Bromo-4-chlorobutyrophenone
Hazard Category H315-H319-H335
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 224-930-3
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only