missing translation for 'onlineSavingsMsg'
Learn More

4-Bromo-4',4″-dimethyltriphenylamine 95.0+%, TCI America™

Catalog No. B30895G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B30895G 5 g
B30891G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B30895G Supplier TCI America Supplier No. B30895G
Only null left

Chemical Identifiers

CAS 58047-42-0
Molecular Formula C20H18BrN
Molecular Weight (g/mol) 352.275
InChI Key YMNJJMJHTXGFOR-UHFFFAOYSA-N
Synonym 4-Bromo-N,N-di-p-tolylaniline
PubChem CID 11142701
IUPAC Name N-(4-bromophenyl)-4-methyl-N-(4-methylphenyl)aniline
SMILES CC1=CC=C(C=C1)N(C2=CC=C(C=C2)C)C3=CC=C(C=C3)Br

Specifications

Melting Point 102°C
Color White-Yellow
Quantity 5 g
Formula Weight 352.28
Percent Purity ≥95.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4-Bromo-4′,4′′-dimethyltriphenylamine