missing translation for 'onlineSavingsMsg'
Learn More

4-Bromo-3-(trifluoromethyl)-1H-pyrazole, 97%, Thermo Scientific™

Catalog No. p-4228473
Change view
Click to view available options
Quantity:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC438040010 1g
AC438040050 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC438040010 Supplier Thermo Scientific Chemicals Supplier No. 438040010

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 19968-17-3
Molecular Formula C4H2BrF3N2
Molecular Weight (g/mol) 214.97
InChI Key JTHNMRUVJDWVMJ-UHFFFAOYSA-N
Synonym 4-bromo-3-trifluoromethyl-1h-pyrazole, 4-bromo-5-trifluoromethyl-1h-pyrazole, 1h-pyrazole, 4-bromo-3-trifluoromethyl, 1h-pyrazole, 4-bromo-5-trifluoromethyl, 1h-pyrazole,4-bromo-3-trifluoromethyl, 4-bromo-3-trifluoromethyl pyrazole, 4-bromo-3-trifluoromethyl-2h-pyrazole, acmc-209f4m, 4-bromo-3 trifluoromethyl-1h-pyrazole
PubChem CID 7147398
IUPAC Name 4-bromo-5-(trifluoromethyl)-1H-pyrazole
SMILES C1=NNC(=C1Br)C(F)(F)F

Specifications

CAS Min % 96.0
CAS Max % 100.0
Infrared Spectrum Authentic
Assay Percent Range 96% min. (HPLC)
Packaging Glass Bottle
Quantity 1g
Formula Weight 214.97
Percent Purity 97%
Chemical Name or Material 4-Bromo-3-(trifluoromethyl)-1H-pyrazole
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
Wear protective gloves/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: If breathing is difficult,remove to fresh air and keep at rest in a position comfortable for breathing.

RUO – Research Use Only