missing translation for 'onlineSavingsMsg'
Learn More

4'-Bromo-3'-nitroacetophenone 98.0+%, TCI America™

Catalog No. p-7122913
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B33831G 1 g
B33835G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B33831G Supplier TCI America Supplier No. B33831G

Chemical Identifiers

CAS 18640-58-9
Molecular Formula C8H6BrNO3
Molecular Weight (g/mol) 244.04
MDL Number MFCD00016985
InChI Key YFVOFFKNHQTQQE-UHFFFAOYSA-N
Synonym 1-4-bromo-3-nitrophenyl ethanone, 4'-bromo-3'-nitroacetophenone, 1-4-bromo-3-nitrophenyl ethan-1-one, 4-bromo-3-nitroacetophenone, 4-bromo-3'-nitroacetophenone, ethanone, 1-4-bromo-3-nitrophenyl, acmc-209eny, 3-nitro-4-bromoacetophenone, 4-bromo-3-nitro acetophenone, 3'-nitro-4'-bromoacetophenone
PubChem CID 87737
IUPAC Name 1-(4-bromo-3-nitrophenyl)ethan-1-one
SMILES CC(=O)C1=CC=C(Br)C(=C1)[N+]([O-])=O

Specifications

Melting Point 119°C
Quantity 1 g
Formula Weight 244.04
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4′-Bromo-3′-nitroacetophenone